Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASZ1
Entrez Gene ID
54897
Gene Name
castor zinc finger 1
Gene Aliases
CAS11, CST, SRG, ZNF693, dJ734G22.1
Chromosome Location
Chr.1: 10636602 - 10796676 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079843.2 NP_001073312.1 18 - 19 4217 55
XM_005263479.3 XP_005263536.1 16 - 17 4081 55
XM_017001539.1 XP_016857028.1 16 - 17 4081 55
XM_017001540.1 XP_016857029.1 18 - 19 4191 55
XM_017001541.1 XP_016857030.1 18 - 19 4253 55
GenBank mRNA AA449398.1 - 2 - 3 306 55

View Full Product Details