Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIP2A
Entrez Gene ID
23181
Gene Name
disco interacting protein 2 homolog A
Gene Aliases
C21orf106, DIP2
Chromosome Location
Chr.21: 46458942 - 46570900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146116.1 NP_001139588.1 27 - 28 3445 79
NM_015151.3 NP_055966.2 27 - 28 3457 79
XM_011529492.2 XP_011527794.1 28 - 29 3572 79
XM_011529501.1 XP_011527803.1 28 - 29 3572 79
XM_017028291.1 XP_016883780.1 28 - 29 3572 79
XM_017028292.1 XP_016883781.1 28 - 29 3572 79
XM_017028293.1 XP_016883782.1 28 - 29 3569 79
XM_017028294.1 XP_016883783.1 27 - 28 3451 79
XM_017028295.1 XP_016883784.1 27 - 28 3448 79
XM_017028296.1 XP_016883785.1 27 - 28 3448 79
XM_017028297.1 XP_016883786.1 27 - 28 3440 79
XM_017028298.1 XP_016883787.1 26 - 27 3319 79
XM_017028299.1 XP_016883788.1 27 - 28 3317 79
XM_017028300.1 XP_016883789.1 28 - 29 3572 79
XM_017028301.1 XP_016883790.1 28 - 29 3572 79
XM_017028302.1 XP_016883791.1 26 - 27 3212 79
XM_017028303.1 XP_016883792.1 28 - 29 3572 79
XM_017028304.1 XP_016883793.1 28 - 29 3572 79
XM_017028305.1 XP_016883794.1 28 - 29 3568 79
GenBank mRNA AB273729.1 - 27 - 28 3295 79
AF490768.1 - 27 - 28 3350 79
AK303351.1 - 25 - 26 3272 79

View Full Product Details