Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPRIN2
Entrez Gene ID
65981
Gene Name
caprin family member 2
Gene Aliases
C1QDC1, EEG-1, EEG1, RNG140
Chromosome Location
Chr.12: 30709552 - 30754994 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002259.2 NP_001002259.1 13 - 14 3042 62
NM_001206856.2 NP_001193785.1 12 - 13 2937 62
NM_001319842.1 NP_001306771.1 12 - 13 2959 62
NM_001319843.1 NP_001306772.1 13 - 14 3042 62
NM_001319844.1 NP_001306773.1 13 - 14 2192 62
NM_001319845.1 NP_001306774.1 14 - 15 2297 62
NM_001319846.1 NP_001306775.1 14 - 15 2287 62
NM_032156.4 NP_115532.3 13 - 14 3042 62
XM_006719138.3 XP_006719201.1 13 - 14 3165 62
XM_006719139.3 XP_006719202.1 12 - 13 3060 62
XM_006719140.3 XP_006719203.1 12 - 13 3060 62
XM_006719145.3 XP_006719208.1 13 - 14 3165 62
XM_006719146.3 XP_006719209.1 13 - 14 3165 62
XM_006719147.3 XP_006719210.1 13 - 14 3165 62
XM_017019851.1 XP_016875340.1 12 - 13 3060 62
XM_017019852.1 XP_016875341.1 14 - 15 2260 62
XM_017019853.1 XP_016875342.1 14 - 15 2330 62
XM_017019855.1 XP_016875344.1 14 - 15 2287 62
XM_017019856.1 XP_016875345.1 13 - 14 2182 62
XM_017019857.1 XP_016875346.1 13 - 14 2155 62
XM_017019858.1 XP_016875347.1 13 - 14 2182 62
XM_017019860.1 XP_016875349.1 13 - 14 3165 62
XM_017019862.1 XP_016875351.1 13 - 14 3165 62
XM_017019863.1 XP_016875352.1 13 - 14 3165 62
XM_017019864.1 XP_016875353.1 13 - 14 3165 62
XM_017019865.1 XP_016875354.1 12 - 13 3060 62
XM_017019866.1 XP_016875355.1 14 - 15 2260 62
XM_017019867.1 XP_016875356.1 14 - 15 2287 62
XM_017019868.1 XP_016875357.1 14 - 15 2260 62
XM_017019869.1 XP_016875358.1 14 - 15 2287 62
XM_017019870.1 XP_016875359.1 14 - 15 2260 62
XM_017019871.1 XP_016875360.1 14 - 15 2331 62
XM_017019872.1 XP_016875361.1 13 - 14 2182 62
XM_017019874.1 XP_016875363.1 13 - 14 2177 62
XM_017019876.1 XP_016875365.1 11 - 12 1788 62
XM_017019877.1 XP_016875366.1 10 - 11 1683 62
XM_017019878.1 XP_016875367.1 10 - 11 1683 62
XM_017019879.1 XP_016875368.1 12 - 13 2072 62
XM_017019881.1 XP_016875370.1 13 - 14 2177 62
XM_017019882.1 XP_016875371.1 13 - 14 2177 62
XM_017019883.1 XP_016875372.1 10 - 11 1683 62
GenBank mRNA AB058776.1 - - 1232 62
AF525416.1 - 5 - 6 553 62
AK021453.1 - 12 - 13 1809 62
AK024247.1 - 1 - 2 646 62
AL834212.1 - 10 - 11 1529 62
AY074491.1 - 12 - 13 2959 62
BC117672.1 - 13 - 14 2839 62
BC117673.1 - 12 - 13 2734 62
BX537569.1 - 11 - 12 3691 62

View Full Product Details