The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014038.2 | NP_054757.1 | 1 - 2 | 155 | 58 |
GenBank mRNA | AF083246.1 | - | 1 - 2 | 155 | 58 |
AK001218.1 | - | 1 - 2 | 135 | 58 | |
AK001260.1 | - | 1 - 2 | 129 | 58 | |
AK300655.1 | - | 1 - 2 | 100 | 58 | |
BC003056.1 | - | 1 - 2 | 81 | 58 | |
BC008453.1 | - | 1 - 2 | 108 | 58 | |
BC009597.1 | - | 1 - 2 | 139 | 58 |