Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABHD13
Entrez Gene ID
84945
Gene Name
abhydrolase domain containing 13
Gene Aliases
BEM46L1, C13orf6, bA153I24.2
Chromosome Location
Chr.13: 108218415 - 108235440 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032859.2 NP_116248.2 2 - 2 2680 141
XM_011521128.2 XP_011519430.1 2 - 2 2638 141
GenBank mRNA AK027812.1 - 1 - 1 2137 141
BX647428.1 - 1 - 1 2260 141

View Full Product Details