The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004984.2 | NP_004975.2 | 4 - 5 | 601 | 75 |
GenBank mRNA | AB210045.1 | - | 4 - 5 | 426 | 75 |
AB290174.1 | - | 4 - 5 | 667 | 75 | |
AK315202.1 | - | 4 - 5 | 621 | 75 | |
BC140009.1 | - | 4 - 5 | 654 | 75 | |
BC146670.1 | - | 4 - 5 | 604 | 75 | |
BC150208.1 | - | 4 - 5 | 604 | 75 | |
U06698.1 | - | 4 - 5 | 541 | 75 |