The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004984.2 | NP_004975.2 | 9 - 10 | 1026 | 76 |
GenBank mRNA | AB210045.1 | - | 9 - 10 | 851 | 76 |
AB290174.1 | - | 9 - 10 | 1092 | 76 | |
AK294881.1 | - | 6 - 7 | 779 | 76 | |
AK315202.1 | - | 9 - 10 | 1046 | 76 | |
BC146670.1 | - | 9 - 10 | 1029 | 76 | |
BC150208.1 | - | 9 - 10 | 1029 | 76 | |
U06698.1 | - | 9 - 10 | 966 | 76 |