The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002660.2 | NP_002651.2 | 26 - 27 | 3253 | 60 |
NM_182811.1 | NP_877963.1 | 26 - 27 | 3253 | 60 | |
XM_005260438.2 | XP_005260495.1 | 26 - 27 | 3255 | 60 | |
XM_011528865.1 | XP_011527167.1 | 26 - 27 | 3120 | 60 | |
GenBank mRNA | AB210028.1 | - | 26 - 27 | 3537 | 60 |
AK297370.1 | - | 15 - 16 | 2003 | 60 | |
AL110247.1 | - | 8 - 9 | 987 | 60 | |
BC136466.1 | - | 26 - 27 | 3220 | 60 | |
BC144136.1 | - | 26 - 27 | 3220 | 60 | |
M34667.1 | - | 26 - 27 | 3208 | 60 |