Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITCH
Entrez Gene ID
83737
Gene Name
itchy E3 ubiquitin protein ligase
Gene Aliases
ADMFD, AIF4, AIP4, NAPP1
Chromosome Location
Chr.20: 34363235 - 34511773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257137.2 NP_001244066.1 20 - 21 2289 115
NM_001257138.2 NP_001244067.1 18 - 19 2024 115
NM_001324197.1 NP_001311126.1 20 - 21 2310 115
NM_001324198.1 NP_001311127.1 19 - 20 2187 115
NM_031483.6 NP_113671.3 19 - 20 2166 115
XM_005260570.1 XP_005260627.1 18 - 19 2082 115
XM_011529079.1 XP_011527381.1 18 - 19 2102 115
XM_017028089.1 XP_016883578.1 19 - 20 2122 115
XM_017028090.1 XP_016883579.1 20 - 21 2242 115
XM_017028091.1 XP_016883580.1 18 - 19 2037 115
GenBank mRNA AB056663.2 - 19 - 20 2076 115
AB209747.1 - 11 - 12 1183 115
AF038564.1 - 14 - 15 1584 115
AF095745.1 - 17 - 18 1959 115
AK297809.1 - 19 - 20 1984 115
AK304090.1 - 18 - 19 1983 115
AK315212.1 - 19 - 20 2125 115
BC011571.1 - 20 - 21 2212 115

View Full Product Details