The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022830.2 | NP_073741.2 | 1 - 2 | 239 | 64 |
GenBank mRNA | AB101208.1 | - | 1 - 2 | 153 | 64 |
AK026000.1 | - | 1 - 2 | 146 | 64 | |
AK292610.1 | - | 1 - 2 | 238 | 64 | |
BC110910.1 | - | 1 - 2 | 128 | 64 | |
BC128263.1 | - | 1 - 2 | 132 | 64 |