The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019593.3 | NP_062539.1 | 8 - 9 | 920 | 97 |
XM_005260758.4 | XP_005260815.1 | 9 - 10 | 1071 | 97 | |
XM_005260761.1 | XP_005260818.1 | 7 - 8 | 844 | 97 | |
XM_006723596.3 | XP_006723659.1 | 9 - 10 | 1844 | 97 | |
GenBank mRNA | AB037855.1 | - | 8 - 9 | 911 | 97 |
AK308111.1 | - | 8 - 9 | 918 | 97 | |
AL833069.1 | - | 3 - 4 | 2116 | 97 | |
BC027588.2 | - | 8 - 9 | 919 | 97 |