The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194946.1 | NP_001181875.1 | 15 - 16 | 2379 | 128 |
NM_001194947.1 | NP_001181876.1 | 14 - 15 | 2322 | 128 | |
NM_001291157.1 | NP_001278086.1 | 14 - 15 | 2411 | 128 | |
NM_198241.2 | NP_937884.1 | 14 - 15 | 2358 | 128 | |
NM_198242.2 | NP_937885.1 | 10 - 11 | 1943 | 128 | |
NM_198244.2 | NP_937887.1 | 11 - 12 | 2120 | 128 | |
GenBank mRNA | AB210013.1 | - | 13 - 14 | 2497 | 128 |
AK096719.1 | - | 12 - 13 | 2008 | 128 | |
AK128378.1 | - | 13 - 14 | 2107 | 128 | |
AK226160.1 | - | 14 - 15 | 2292 | 128 | |
AK294567.1 | - | 12 - 13 | 1989 | 128 | |
AK299763.1 | - | 11 - 12 | 2048 | 128 | |
AK308319.1 | - | 9 - 10 | 1708 | 128 | |
AL832754.1 | - | 10 - 11 | 1832 | 128 | |
BC082764.1 | - | 7 - 8 | 1516 | 128 | |
BC140892.1 | - | 15 - 16 | 2221 | 128 | |
BC140896.1 | - | 14 - 15 | 2164 | 128 | |
BX647812.1 | - | 14 - 15 | 2271 | 128 |