The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001941.4 | NP_001932.2 | 13 - 14 | 2262 | 131 |
NM_024423.3 | NP_077741.2 | 13 - 14 | 2262 | 131 | |
GenBank mRNA | AF297090.1 | - | 2 - 3 | 369 | 131 |
AK291748.1 | - | 13 - 14 | 2188 | 131 | |
BG697535.1 | - | 1 - 2 | 231 | 131 | |
D17427.1 | - | 13 - 14 | 2174 | 131 | |
X83929.1 | - | 14 - 15 | 2128 | 131 |