Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DSC3
Entrez Gene ID
1825
Gene Name
desmocollin 3
Gene Aliases
CDHF3, DSC, DSC1, DSC2, DSC4, HT-CP
Chromosome Location
Chr.18: 30989365 - 31042815 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024423.3 NP_077741.2 16 - 17 2694 81

View Full Product Details