Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DSC3
Entrez Gene ID
1825
Gene Name
desmocollin 3
Gene Aliases
CDHF3, DSC, DSC1, DSC2, DSC4, HT-CP
Chromosome Location
Chr.18: 30989365 - 31042815 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001941.4 NP_001932.2 2 - 3 306 128
NM_024423.3 NP_077741.2 2 - 3 306 128
GenBank mRNA AK291748.1 - 2 - 3 232 128
D17427.1 - 2 - 3 218 128
DA860471.1 - 2 - 3 306 128
X83929.1 - 3 - 4 172 128

View Full Product Details