Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DSC3
Entrez Gene ID
1825
Gene Name
desmocollin 3
Gene Aliases
CDHF3, DSC, DSC1, DSC2, DSC4, HT-CP
Chromosome Location
Chr.18: 30989365 - 31042815 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001941.4 NP_001932.2 3 - 4 511 77
NM_024423.3 NP_077741.2 3 - 4 511 77
GenBank mRNA AK291748.1 - 3 - 4 437 77
D17427.1 - 3 - 4 423 77
X83929.1 - 4 - 5 377 77

View Full Product Details