Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENTPD7
Entrez Gene ID
57089
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1
Chromosome Location
Chr.10: 99659506 - 99711245 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020354.3 NP_065087.1 1 - 2 86 64
GenBank mRNA AF269255.1 - 1 - 2 51 64
AK055540.1 - 1 - 2 86 64
AK309735.1 - 1 - 2 86 64
AK314542.1 - 1 - 2 144 64
BC119008.1 - 1 - 2 52 64
BC122857.1 - 1 - 2 52 64

View Full Product Details