Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENTPD7
Entrez Gene ID
57089
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1
Chromosome Location
Chr.10: 99659506 - 99711245 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020354.3 NP_065087.1 3 - 4 373 91
GenBank mRNA AF269255.1 - 3 - 4 338 91
AK055540.1 - 3 - 4 373 91
AK309735.1 - 4 - 5 556 91
AK314542.1 - 3 - 4 431 91
BC119008.1 - 3 - 4 339 91
BC122857.1 - 3 - 4 339 91

View Full Product Details