The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020354.3 | NP_065087.1 | 5 - 6 | 729 | 105 |
GenBank mRNA | AF269255.1 | - | 5 - 6 | 694 | 105 |
AK055540.1 | - | 5 - 6 | 729 | 105 | |
AK309735.1 | - | 6 - 7 | 912 | 105 | |
AK314542.1 | - | 5 - 6 | 787 | 105 | |
BC119008.1 | - | 5 - 6 | 695 | 105 | |
BC122857.1 | - | 5 - 6 | 695 | 105 |