Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIRT1
Entrez Gene ID
23411
Gene Name
sirtuin 1
Gene Aliases
SIR2, SIR2L1, SIR2alpha
Chromosome Location
Chr.10: 67884669 - 67918390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142498.1 NP_001135970.1 3 - 4 488 109
NM_012238.4 NP_036370.2 4 - 5 994 109
GenBank mRNA AF083106.2 - 4 - 5 994 109
AF235040.1 - 4 - 5 989 109
AK289743.1 - 4 - 5 730 109
AL136741.1 - 4 - 5 609 109
BC012499.1 - 4 - 5 574 109
DB162635.1 - 3 - 4 485 109
JQ768366.1 - 2 - 3 365 109

View Full Product Details