Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PACRGL
Entrez Gene ID
133015
Gene Name
PARK2 coregulated like
Gene Aliases
C4orf28
Chromosome Location
Chr.4: 20696282 - 20757766 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258345.1 NP_001245274.1 5 - 6 757 139
NM_145048.3 NP_659485.1 5 - 6 757 139
NR_133933.1 5 - 6 614 139
XM_011513780.1 XP_011512082.1 7 - 8 920 139
XM_011513781.1 XP_011512083.1 7 - 8 739 139
XM_011513784.1 XP_011512086.1 7 - 8 756 139
XM_011513785.1 XP_011512087.1 6 - 7 839 139
XM_011513786.2 XP_011512088.1 7 - 8 920 139
XM_011513787.1 XP_011512089.1 5 - 6 711 139
XM_011513788.1 XP_011512090.1 6 - 7 828 139
XM_011513791.1 XP_011512093.1 5 - 6 671 139
XM_011513792.1 XP_011512094.1 5 - 6 666 139
XM_011513794.1 XP_011512096.1 6 - 7 619 139
XM_011513795.1 XP_011512097.1 5 - 6 541 139
XM_011513796.1 XP_011512098.1 5 - 6 536 139
XM_011513797.2 XP_011512099.1 5 - 6 542 139
XM_011513798.2 XP_011512100.1 5 - 6 478 139
XM_011513805.2 XP_011512107.1 5 - 6 542 139
XM_017007747.1 XP_016863236.1 7 - 8 756 139
XM_017007748.1 XP_016863237.1 6 - 7 626 139
XM_017007749.1 XP_016863238.1 5 - 6 541 139
XM_017007751.1 XP_016863240.1 6 - 7 824 139
XM_017007752.1 XP_016863241.1 5 - 6 671 139
XM_017007753.1 XP_016863242.1 5 - 6 541 139
GenBank mRNA AK098692.1 - 5 - 6 614 139
AK291544.1 - 5 - 6 713 139
AK307092.1 - 4 - 5 460 139
AK307786.1 - 7 - 8 756 139
AK315482.1 - 5 - 6 712 139
BC023002.2 - 5 - 6 504 139

View Full Product Details