The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001890.1 | NP_001881.1 | 11 - 12 | 345 | 80 |
XM_006714089.2 | XP_006714152.1 | 11 - 12 | 361 | 80 | |
XM_006714090.2 | XP_006714153.1 | 10 - 11 | 340 | 80 | |
XM_006714091.2 | XP_006714154.1 | 10 - 11 | 337 | 80 | |
GenBank mRNA | BC128227.1 | - | 11 - 12 | 334 | 80 |
BC128228.1 | - | 11 - 12 | 334 | 80 | |
U23157.1 | - | 11 - 12 | 324 | 80 | |
X78416.1 | - | 11 - 12 | 345 | 80 | |
X98084.1 | - | 10 - 11 | 273 | 80 |