The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083907.2 | NP_001077376.2 | 9 - 10 | 1632 | 81 |
NM_001127507.2 | NP_001120979.2 | 8 - 9 | 1464 | 81 | |
NM_017935.4 | NP_060405.4 | 9 - 10 | 1863 | 81 | |
XM_017008337.1 | XP_016863826.1 | 9 - 10 | 1610 | 81 | |
GenBank mRNA | AB063170.1 | - | 8 - 9 | 1499 | 81 |
AK000713.1 | - | 9 - 10 | 1579 | 81 | |
AK091523.1 | - | 13 - 14 | 1964 | 81 | |
AK292133.1 | - | 9 - 10 | 1707 | 81 | |
AL834291.1 | - | 9 - 10 | 1682 | 81 | |
BC032241.1 | - | 9 - 10 | 1731 | 81 | |
EU051376.1 | - | 8 - 9 | 1202 | 81 |