Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECT2L
Entrez Gene ID
345930
Gene Name
epithelial cell transforming 2 like
Gene Aliases
ARHGEF32, C6orf91, FBXO49, LFDH, dJ509I19.2, dJ509I19.3, dJ509I19.5
Chromosome Location
Chr.6: 138796107 - 138904070 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077706.2 NP_001071174.1 5 - 6 663 61
NM_001195037.2 NP_001181966.1 4 - 5 523 61
XM_006715472.3 XP_006715535.1 5 - 6 656 61
XM_011535795.2 XP_011534097.1 4 - 5 527 61
XM_011535797.2 XP_011534099.1 2 - 3 239 61
XM_017010828.1 XP_016866317.1 5 - 6 629 61
XM_017010829.1 XP_016866318.1 5 - 6 656 61
XM_017010830.1 XP_016866319.1 5 - 6 656 61
GenBank mRNA AK304425.1 - 2 - 3 221 61
BC146884.1 - 3 - 4 370 61
DC418522.1 - 4 - 5 522 61
DQ904331.1 - 3 - 4 317 61

View Full Product Details