The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143850.2 | NP_001137322.1 | 2 - 3 | 314 | 139 |
NM_024809.4 | NP_079085.2 | 2 - 3 | 314 | 139 | |
XM_005253623.2 | XP_005253680.1 | 2 - 3 | 314 | 139 | |
XM_006719605.3 | XP_006719668.1 | 2 - 3 | 314 | 139 | |
XM_017019974.1 | XP_016875463.1 | 2 - 3 | 314 | 139 | |
XM_017019975.1 | XP_016875464.1 | 2 - 3 | 319 | 139 | |
GenBank mRNA | AK056924.1 | - | 2 - 3 | 312 | 139 |
AK292153.1 | - | 2 - 3 | 314 | 139 | |
BC009112.1 | - | 2 - 3 | 283 | 139 | |
DQ278870.1 | - | 2 - 3 | 312 | 139 |