The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271842.1 | NP_001258771.1 | 3 - 4 | 283 | 74 |
NM_001319190.1 | NP_001306119.1 | 3 - 4 | 283 | 74 | |
NM_207103.3 | NP_996986.1 | 3 - 4 | 304 | 74 | |
XM_017024615.1 | XP_016880104.1 | 3 - 4 | 283 | 74 | |
GenBank mRNA | AK057142.1 | - | 3 - 4 | 246 | 74 |
AK297035.1 | - | 3 - 4 | 231 | 74 | |
AK310145.1 | - | 3 - 4 | 283 | 74 | |
AY358809.1 | - | 3 - 4 | 286 | 74 | |
BC127925.1 | - | 3 - 4 | 239 | 74 | |
BC130554.1 | - | 3 - 4 | 232 | 74 | |
BC130556.1 | - | 3 - 4 | 232 | 74 | |
BC144395.1 | - | 3 - 4 | 211 | 74 | |
DQ778084.1 | - | 3 - 4 | 178 | 74 |