The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002266.2 | NP_001002266.1 | 5 - 6 | 686 | 70 |
NM_001282866.1 | NP_001269795.1 | 6 - 7 | 1532 | 70 | |
NM_145021.5 | NP_659458.2 | 5 - 6 | 1174 | 70 | |
XM_005271804.2 | XP_005271861.1 | 6 - 7 | 2021 | 70 | |
XM_006717704.2 | XP_006717767.1 | 6 - 7 | 1436 | 70 | |
XM_011539492.2 | XP_011537794.1 | 7 - 8 | 2072 | 70 | |
XM_011539493.2 | XP_011537795.1 | 8 - 9 | 2313 | 70 | |
XM_011539494.2 | XP_011537796.1 | 7 - 8 | 2262 | 70 | |
XM_011539495.1 | XP_011537797.1 | 5 - 6 | 570 | 70 | |
XM_017015894.1 | XP_016871383.1 | 6 - 7 | 1226 | 70 | |
GenBank mRNA | AK023636.1 | - | 1 - 2 | 975 | 70 |
AK307667.1 | - | 6 - 7 | 1532 | 70 | |
AK313340.1 | - | 5 - 6 | 643 | 70 | |
AL833316.1 | - | 2 - 3 | 917 | 70 | |
BC025394.2 | - | 5 - 6 | 663 | 70 | |
BC066988.1 | - | 5 - 6 | 1175 | 70 | |
BG433948.1 | - | 2 - 3 | 227 | 70 |