The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013703.3 | NP_001013725.2 | 13 - 14 | 2388 | 104 |
XM_005254392.2 | XP_005254449.1 | 13 - 14 | 2411 | 104 | |
XM_011521599.2 | XP_011519901.1 | 13 - 14 | 2411 | 104 | |
XM_011521600.2 | XP_011519902.1 | 13 - 14 | 2411 | 104 | |
XM_017022219.1 | XP_016877708.1 | 13 - 14 | 2411 | 104 | |
GenBank mRNA | AB037759.1 | - | 9 - 10 | 1854 | 104 |
AL157497.1 | - | 2 - 3 | 185 | 104 |