The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013703.3 | NP_001013725.2 | 24 - 25 | 3485 | 85 |
XM_005254392.2 | XP_005254449.1 | 24 - 25 | 3508 | 85 | |
XM_011521599.2 | XP_011519901.1 | 24 - 25 | 3508 | 85 | |
XM_011521600.2 | XP_011519902.1 | 24 - 25 | 3508 | 85 | |
XM_017022219.1 | XP_016877708.1 | 24 - 25 | 3508 | 85 | |
GenBank mRNA | AB037759.1 | - | 20 - 21 | 2951 | 85 |
AJ243428.1 | - | 2 - 3 | 112 | 85 | |
AK027011.1 | - | 10 - 11 | 923 | 85 | |
AK074361.1 | - | 3 - 4 | 324 | 85 | |
AL157497.1 | - | 13 - 14 | 1282 | 85 | |
AL832907.1 | - | 18 - 19 | 2745 | 85 | |
BC009350.2 | - | 10 - 11 | 955 | 85 | |
BX649048.1 | - | 7 - 8 | 2330 | 85 |