The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013703.3 | NP_001013725.2 | 36 - 37 | 4805 | 108 |
XM_011521600.2 | XP_011519902.1 | 35 - 36 | 4657 | 108 | |
GenBank mRNA | AB037759.1 | - | 32 - 33 | 4271 | 108 |
AJ243428.1 | - | 14 - 15 | 1432 | 108 | |
AK027011.1 | - | 22 - 23 | 2243 | 108 | |
AK074361.1 | - | 14 - 15 | 1473 | 108 | |
AL137627.1 | - | 5 - 6 | 359 | 108 | |
AL157497.1 | - | 25 - 26 | 2602 | 108 | |
AL832907.1 | - | 30 - 31 | 4065 | 108 | |
BC009350.2 | - | 22 - 23 | 2275 | 108 |