The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013703.3 | NP_001013725.2 | 8 - 9 | 1090 | 106 |
XM_005254392.2 | XP_005254449.1 | 8 - 9 | 1113 | 106 | |
XM_011521599.2 | XP_011519901.1 | 8 - 9 | 1113 | 106 | |
XM_011521600.2 | XP_011519902.1 | 8 - 9 | 1113 | 106 | |
XM_017022219.1 | XP_016877708.1 | 8 - 9 | 1113 | 106 | |
GenBank mRNA | AB037759.1 | - | 4 - 5 | 556 | 106 |
AL832907.1 | - | 3 - 4 | 434 | 106 | |
BC078179.1 | - | 8 - 9 | 1068 | 106 |