Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED13L
Entrez Gene ID
23389
Gene Name
mediator complex subunit 13 like
Gene Aliases
MRFACD, PROSIT240, THRAP2, TRAP240L
Chromosome Location
Chr.12: 115958576 - 116277219 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015335.4 NP_056150.1 14 - 15 2627 69
XM_011538080.2 XP_011536382.1 14 - 15 2660 69
XM_011538082.2 XP_011536384.1 14 - 15 2830 69
GenBank mRNA AB028948.2 - 8 - 9 1694 69
AF515599.1 - 14 - 15 2627 69
AK127706.1 - 11 - 12 2161 69
AL133033.1 - 8 - 9 1667 69
AY338463.1 - 14 - 15 2779 69
BC130422.1 - 14 - 15 2623 69

View Full Product Details