The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015335.4 | NP_056150.1 | 16 - 17 | 3051 | 94 |
XM_011538080.2 | XP_011536382.1 | 16 - 17 | 3084 | 94 | |
XM_011538081.2 | XP_011536383.1 | 16 - 17 | 3081 | 94 | |
XM_011538082.2 | XP_011536384.1 | 16 - 17 | 3254 | 94 | |
XM_017019090.1 | XP_016874579.1 | 16 - 17 | 3081 | 94 | |
GenBank mRNA | AB028948.2 | - | 10 - 11 | 2118 | 94 |
AF515599.1 | - | 16 - 17 | 3051 | 94 | |
AK127706.1 | - | 13 - 14 | 2585 | 94 | |
AL133033.1 | - | 10 - 11 | 2091 | 94 | |
AY338463.1 | - | 16 - 17 | 3203 | 94 | |
BC130422.1 | - | 16 - 17 | 3047 | 94 | |
CR749332.1 | - | 14 - 15 | 2575 | 94 |