The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144628.3 | NP_653229.1 | 6 - 7 | 915 | 59 |
NR_111901.1 | 6 - 7 | 915 | 59 | ||
XM_005260661.1 | XP_005260718.1 | 6 - 7 | 905 | 59 | |
XM_006723540.3 | XP_006723603.1 | 5 - 6 | 694 | 59 | |
XM_017027645.1 | XP_016883134.1 | 5 - 6 | 686 | 59 | |
GenBank mRNA | AB449906.1 | - | 6 - 7 | 767 | 59 |
AK055573.1 | - | 1 - 2 | 1394 | 59 | |
AK095110.1 | - | 5 - 6 | 1074 | 59 | |
AK127062.1 | - | 6 - 7 | 895 | 59 | |
AK291648.1 | - | 6 - 7 | 878 | 59 | |
BC014983.1 | - | 6 - 7 | 865 | 59 | |
BX647370.1 | - | 1 - 2 | 1645 | 59 | |
DA230581.1 | - | 3 - 4 | 428 | 59 |