Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX20
Entrez Gene ID
11218
Gene Name
DEAD-box helicase 20
Gene Aliases
DP103, GEMIN3
Chromosome Location
Chr.1: 111755568 - 111767577 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007204.4 NP_009135.4 3 - 4 923 104
GenBank mRNA AF106019.1 - 3 - 4 576 104
AF155803.1 - 3 - 4 571 104
AF171063.1 - 3 - 4 859 104
AK001506.1 - 3 - 4 585 104
AK314312.1 - 3 - 4 568 104
BC011556.1 - 3 - 4 576 104
BC031062.1 - 3 - 4 585 104
BC034953.2 - 3 - 4 894 104

View Full Product Details