The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005430.3 | NP_005421.1 | 3 - 4 | 823 | 67 |
GenBank mRNA | BC074798.2 | - | 3 - 4 | 661 | 67 |
BC074799.2 | - | 3 - 4 | 661 | 67 | |
BT019429.1 | - | 3 - 4 | 625 | 67 |