The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017969.2 | NP_060439.2 | 3 - 4 | 1518 | 123 |
XM_005263707.3 | XP_005263764.1 | 3 - 4 | 1354 | 123 | |
XM_006712626.3 | XP_006712689.1 | 3 - 4 | 1432 | 123 | |
XM_011511450.1 | XP_011509752.1 | 3 - 4 | 1432 | 123 | |
XM_011511452.1 | XP_011509754.1 | 3 - 4 | 1432 | 123 | |
XM_017004467.1 | XP_016859956.1 | 3 - 4 | 1519 | 123 | |
XM_017004468.1 | XP_016859957.1 | 3 - 4 | 1288 | 123 | |
GenBank mRNA | AK000868.1 | - | 3 - 4 | 845 | 123 |
AK027561.1 | - | 3 - 4 | 1475 | 123 | |
AK056881.1 | - | 3 - 4 | 519 | 123 | |
AK294669.1 | - | 4 - 5 | 1226 | 123 | |
AK295970.1 | - | 3 - 4 | 1281 | 123 | |
AK296848.1 | - | 4 - 5 | 1326 | 123 | |
AL834178.1 | - | 3 - 4 | 1466 | 123 | |
BC110536.1 | - | 3 - 4 | 1348 | 123 | |
BC110537.1 | - | 3 - 4 | 1348 | 123 |