The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244713.1 | NP_001231642.1 | 3 - 4 | 308 | 131 |
NM_001244724.1 | NP_001231653.1 | 3 - 4 | 298 | 131 | |
NM_002874.4 | NP_002865.1 | 3 - 4 | 623 | 131 | |
GenBank mRNA | AK125226.1 | - | 4 - 5 | 527 | 131 |
AK223479.1 | - | 3 - 4 | 575 | 131 | |
AK293532.1 | - | 3 - 4 | 615 | 131 | |
AK297986.1 | - | 3 - 4 | 268 | 131 | |
AK316189.1 | - | 3 - 4 | 223 | 131 | |
AL540969.3 | - | 3 - 4 | 594 | 131 | |
AY313777.1 | - | 3 - 4 | 298 | 131 | |
BC020973.2 | - | 3 - 4 | 279 | 131 | |
BI460482.1 | - | 3 - 4 | 315 | 131 | |
D21090.1 | - | 3 - 4 | 542 | 131 |