The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005188.3 | NP_005179.2 | 3 - 4 | 732 | 79 |
XM_011543057.2 | XP_011541359.1 | 3 - 4 | 732 | 79 | |
XM_017018511.1 | XP_016874000.1 | 2 - 3 | 167 | 79 | |
GenBank mRNA | BC132733.1 | - | 3 - 4 | 661 | 79 |
BC136463.1 | - | 3 - 4 | 661 | 79 | |
X57110.1 | - | 3 - 4 | 738 | 79 |