The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005188.3 | NP_005179.2 | 7 - 8 | 1233 | 69 |
XM_011543057.2 | XP_011541359.1 | 7 - 8 | 1233 | 69 | |
XM_017018511.1 | XP_016874000.1 | 6 - 7 | 668 | 69 | |
GenBank mRNA | BC132733.1 | - | 7 - 8 | 1162 | 69 |
BC136463.1 | - | 7 - 8 | 1162 | 69 | |
X57110.1 | - | 7 - 8 | 1239 | 69 |