The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013270.4 | NP_037402.1 | 3 - 4 | 533 | 69 |
GenBank mRNA | AF100707.1 | - | 3 - 4 | 533 | 69 |
BC033016.1 | - | 3 - 4 | 524 | 69 | |
BC037775.2 | - | 3 - 4 | 495 | 69 |