Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRRM1
Entrez Gene ID
10250
Gene Name
serine and arginine repetitive matrix 1
Gene Aliases
160-KD, POP101, SRM160
Chromosome Location
Chr.1: 24643012 - 24673281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303448.1 NP_001290377.1 12 - 13 1845 78
NM_001303449.1 NP_001290378.1 12 - 13 1774 78
XM_005245717.2 XP_005245774.1 12 - 13 1761 78
XM_005245718.2 XP_005245775.1 12 - 13 1758 78
XM_005245720.2 XP_005245777.1 12 - 13 1752 78
XM_005245722.2 XP_005245779.1 12 - 13 1671 78
XM_011540478.2 XP_011538780.1 12 - 13 1775 78
XM_011540479.1 XP_011538781.1 12 - 13 3722 78
XM_011540480.1 XP_011538782.1 13 - 14 3030 78
XM_011540481.1 XP_011538783.1 13 - 14 2072 78
XM_011540482.1 XP_011538784.1 12 - 13 3716 78
XM_011540483.1 XP_011538785.1 12 - 13 3632 78
XM_017000011.1 XP_016855500.1 12 - 13 1772 78
XM_017000012.1 XP_016855501.1 12 - 13 1766 78
XM_017000016.1 XP_016855505.1 12 - 13 1707 78
XM_017000017.1 XP_016855506.1 12 - 13 1682 78
XM_017000020.1 XP_016855509.1 12 - 13 1668 78
XM_017000021.1 XP_016855510.1 12 - 13 1665 78
XM_017000023.1 XP_016855512.1 12 - 13 3719 78
XM_017000024.1 XP_016855513.1 12 - 13 3713 78
XM_017000030.1 XP_016855519.1 12 - 13 3629 78
XM_017000031.1 XP_016855520.1 13 - 14 1979 78
XM_017000032.1 XP_016855521.1 12 - 13 3626 78
GenBank mRNA AK293576.1 - 10 - 11 1268 78
AK302498.1 - 12 - 13 1664 78
AK307419.1 - 12 - 13 1774 78
AK307568.1 - 9 - 10 1485 78
KU178513.1 - 12 - 13 1627 78

View Full Product Details