The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017581.3 | NP_060051.2 | 2 - 3 | 348 | 74 |
GenBank mRNA | AF227732.1 | - | 2 - 3 | 191 | 74 |
AJ243342.1 | - | 2 - 3 | 225 | 74 | |
BC113549.1 | - | 2 - 3 | 304 | 74 | |
BC113575.1 | - | 2 - 3 | 304 | 74 |