The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128204.1 | NP_001121676.1 | 11 - 12 | 1803 | 90 |
NM_001128205.1 | NP_001121677.1 | 12 - 13 | 1971 | 90 | |
NM_001128206.1 | NP_001121678.1 | 11 - 12 | 1809 | 90 | |
NM_015170.2 | NP_055985.2 | 12 - 13 | 1965 | 90 | |
NR_132437.1 | 5 - 6 | 598 | 90 | ||
XM_006716438.2 | XP_006716501.1 | 12 - 13 | 2003 | 90 | |
XM_006716439.2 | XP_006716502.1 | 11 - 12 | 1809 | 90 | |
XM_006716440.3 | XP_006716503.1 | 12 - 13 | 1965 | 90 | |
XM_006716441.1 | XP_006716504.1 | 5 - 6 | 598 | 90 | |
XM_006716442.1 | XP_006716505.1 | 5 - 6 | 598 | 90 | |
XM_011517494.1 | XP_011515796.1 | 11 - 12 | 1714 | 90 | |
XM_011517495.1 | XP_011515797.1 | 12 - 13 | 2003 | 90 | |
XM_017013250.1 | XP_016868739.1 | 12 - 13 | 2003 | 90 | |
GenBank mRNA | AB029000.1 | - | 7 - 8 | 1089 | 90 |
AF545571.1 | - | 12 - 13 | 1954 | 90 | |
AK055607.1 | - | 11 - 12 | 1753 | 90 | |
AK074873.1 | - | 3 - 4 | 325 | 90 | |
AK095341.1 | - | 11 - 12 | 1804 | 90 | |
AK307951.1 | - | 11 - 12 | 1583 | 90 | |
AY101175.1 | - | 11 - 12 | 1756 | 90 | |
BC068565.1 | - | 5 - 6 | 588 | 90 | |
BX571746.1 | - | 12 - 13 | 1916 | 90 |