Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SULF1
Entrez Gene ID
23213
Gene Name
sulfatase 1
Gene Aliases
SULF-1
Chromosome Location
Chr.8: 69466624 - 69660912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128204.1 NP_001121676.1 17 - 18 2664 136
NM_001128205.1 NP_001121677.1 18 - 19 2832 136
NM_001128206.1 NP_001121678.1 17 - 18 2670 136
NM_015170.2 NP_055985.2 18 - 19 2826 136
NR_132437.1 11 - 12 1459 136
XM_006716438.2 XP_006716501.1 18 - 19 2864 136
XM_006716439.2 XP_006716502.1 17 - 18 2670 136
XM_006716440.3 XP_006716503.1 18 - 19 2826 136
XM_006716441.1 XP_006716504.1 11 - 12 1459 136
XM_006716442.1 XP_006716505.1 11 - 12 1459 136
XM_011517494.1 XP_011515796.1 17 - 18 2575 136
XM_011517495.1 XP_011515797.1 18 - 19 2864 136
XM_017013250.1 XP_016868739.1 18 - 19 2864 136
GenBank mRNA AB029000.1 - 13 - 14 1950 136
AF545571.1 - 18 - 19 2815 136
AK074873.1 - 9 - 10 1186 136
AK123744.1 - 2 - 3 130 136
AK307951.1 - 17 - 18 2444 136
AY101175.1 - 17 - 18 2617 136
BC068565.1 - 11 - 12 1449 136
BX571746.1 - 18 - 19 2777 136

View Full Product Details