The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173854.5 | NP_776253.3 | 4 - 5 | 1590 | 72 |
XM_005245069.2 | XP_005245126.1 | 4 - 5 | 1266 | 72 | |
GenBank mRNA | AJ514402.1 | - | 3 - 4 | 677 | 72 |
AK128069.1 | - | 3 - 4 | 843 | 72 | |
AK314732.1 | - | 3 - 4 | 748 | 72 | |
AL831974.1 | - | 4 - 5 | 1426 | 72 | |
AL832001.1 | - | 4 - 5 | 1426 | 72 | |
BC136303.1 | - | 3 - 4 | 1071 | 72 | |
BX641054.1 | - | 4 - 5 | 1428 | 72 | |
BX648979.1 | - | 4 - 5 | 1464 | 72 |