The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000466.2 | NP_000457.1 | 11 - 12 | 1997 | 81 |
NM_001282678.1 | NP_001269607.1 | 11 - 12 | 2032 | 81 | |
XM_017012319.1 | XP_016867808.1 | 6 - 7 | 917 | 81 | |
GenBank mRNA | AB008112.1 | - | 11 - 12 | 1968 | 81 |
AB052091.1 | - | 11 - 12 | 1901 | 81 | |
AB052092.1 | - | 11 - 12 | 1901 | 81 | |
AB052094.1 | - | 11 - 12 | 1901 | 81 | |
AB209355.1 | - | 1 - 2 | 1577 | 81 | |
AF026086.1 | - | 11 - 12 | 1961 | 81 | |
AF030356.1 | - | 11 - 12 | 1905 | 81 | |
AK292955.1 | - | 11 - 12 | 1902 | 81 | |
AK293757.1 | - | 11 - 12 | 1940 | 81 | |
AK293848.1 | - | 6 - 7 | 719 | 81 | |
AK295686.1 | - | 8 - 9 | 941 | 81 | |
BC035575.1 | - | 11 - 12 | 1939 | 81 |