The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000466.2 | NP_000457.1 | 17 - 18 | 2886 | 103 |
NM_001282677.1 | NP_001269606.1 | 16 - 17 | 2715 | 103 | |
NM_001282678.1 | NP_001269607.1 | 17 - 18 | 2921 | 103 | |
XM_017012319.1 | XP_016867808.1 | 12 - 13 | 1806 | 103 | |
GenBank mRNA | AB008112.1 | - | 17 - 18 | 2857 | 103 |
AB052090.1 | - | 16 - 17 | 2619 | 103 | |
AB052091.1 | - | 17 - 18 | 2790 | 103 | |
AB052092.1 | - | 17 - 18 | 2790 | 103 | |
AB052093.1 | - | 17 - 18 | 2790 | 103 | |
AB052094.1 | - | 17 - 18 | 2790 | 103 | |
AB209355.1 | - | 6 - 7 | 2682 | 103 | |
AF026086.1 | - | 17 - 18 | 2850 | 103 | |
AF030356.1 | - | 17 - 18 | 2794 | 103 | |
AK292955.1 | - | 17 - 18 | 2791 | 103 | |
AK293757.1 | - | 17 - 18 | 2829 | 103 | |
AK295686.1 | - | 14 - 15 | 1830 | 103 | |
BC035575.1 | - | 17 - 18 | 2828 | 103 |