The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000466.2 | NP_000457.1 | 4 - 5 | 569 | 155 |
NM_001282677.1 | NP_001269606.1 | 4 - 5 | 569 | 155 | |
NM_001282678.1 | NP_001269607.1 | 4 - 5 | 604 | 155 | |
GenBank mRNA | AB008112.1 | - | 4 - 5 | 540 | 155 |
AB052090.1 | - | 4 - 5 | 473 | 155 | |
AB052091.1 | - | 4 - 5 | 473 | 155 | |
AB052092.1 | - | 4 - 5 | 473 | 155 | |
AB052093.1 | - | 4 - 5 | 473 | 155 | |
AB052094.1 | - | 4 - 5 | 473 | 155 | |
AF026086.1 | - | 4 - 5 | 533 | 155 | |
AF030356.1 | - | 4 - 5 | 477 | 155 | |
AK292955.1 | - | 4 - 5 | 474 | 155 | |
AK293757.1 | - | 4 - 5 | 512 | 155 | |
BC035575.1 | - | 4 - 5 | 511 | 155 | |
DC370317.1 | - | 4 - 5 | 580 | 155 |