Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TCTA
Entrez Gene ID
6988
Gene Name
T-cell leukemia translocation altered
Gene Aliases
-
Chromosome Location
Chr.3: 49412206 - 49416476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022171.2 NP_071503.1 1 - 2 438 65
GenBank mRNA AK000824.1 - 1 - 2 255 65
AK311839.1 - 1 - 2 221 65
BC005157.1 - 1 - 2 404 65
CR457411.1 - 1 - 2 217 65
L41143.1 - 1 - 2 438 65

View Full Product Details